Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164443.1 | NP_001157915.1 | 7 | 699 | ||
XM_011543298.2 | XP_011541600.1 | 7 | 721 | |||
XM_011543299.1 | XP_011541601.1 | 7 | 699 | |||
XM_011543300.2 | XP_011541602.1 | 7 | 721 | |||
XM_011543301.2 | XP_011541603.1 | 7 | 721 | |||
XM_011543302.2 | XP_011541604.1 | 5 | 257 | |||
XM_017009317.1 | XP_016864806.1 | 7 | 616 | |||
XM_017009318.1 | XP_016864807.1 | 7 | 721 | |||
XM_017009319.1 | XP_016864808.1 | 7 | 721 | |||
XM_017009320.1 | XP_016864809.1 | 4 | 242 | |||
XM_017009322.1 | XP_016864811.1 | 7 | 721 | |||
GenBank mRNA | AK097510.1 | - | 7 | 699 |