Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152774.3 | NP_689987.3 | 4 | 561 | ||
XM_005249692.4 | XP_005249749.2 | 5 | 1572 | |||
XM_011515249.2 | XP_011513551.1 | 5 | 1554 | |||
XM_011515250.2 | XP_011513552.1 | 4 | 1498 | |||
GenBank mRNA | BC030104.1 | - | 5 | 433 | 4797383 | |
BX647256.1 | - | 4 | 1172 | |||
BX648384.1 | - | 4 | 562 | |||
DN991960.1 | - | 3 | 342 |