Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318380.1 | NP_001305309.1 | 2, 3 | 239 | ||
NM_177533.4 | NP_803877.2 | 2, 3 | 239 | |||
GenBank mRNA | AB087802.1 | - | 2, 3 | 125 | ||
AK128196.1 | - | 2, 3 | 228 | |||
AK310449.1 | - | 2, 3 | 216 | |||
BC041584.1 | - | 2, 3 | 201 | 4810207 | ||
DA249638.1 | - | 2, 3 | 159 | |||
DA409636.1 | - | 2, 3 | 222 | |||
DB096906.1 | - | 2, 3 | 241 |