Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NATD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152914.2 | NP_690878.2 | 3 | 285 | ||
GenBank mRNA | AK126856.1 | - | 3 | 285 | ||
BC029527.1 | - | 3 | 429 | 5302828 |