Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286408.1 | NP_001273337.1 | 2 | 212 | ||
NM_198081.4 | NP_932347.2 | 3 | 427 | |||
XM_011535703.2 | XP_011534005.1 | 3 | 325 | |||
XM_011535704.2 | XP_011534006.1 | 3 | 203 | |||
XM_011535706.1 | XP_011534008.1 | 2 | 169 | |||
XM_011535707.2 | XP_011534009.1 | 3 | 446 | |||
XM_017010677.1 | XP_016866166.1 | 3 | 494 | |||
XM_017010678.1 | XP_016866167.1 | 3 | 407 | |||
XM_017010679.1 | XP_016866168.1 | 3 | 305 | |||
XM_017010680.1 | XP_016866169.1 | 3 | 373 | |||
XM_017010681.1 | XP_016866170.1 | 3 | 283 | |||
XM_017010682.1 | XP_016866171.1 | 3 | 497 | |||
XM_017010683.1 | XP_016866172.1 | 2 | 166 | |||
XM_017010684.1 | XP_016866173.1 | 3 | 499 | |||
XM_017010685.1 | XP_016866174.1 | 3 | 494 | |||
XM_017010686.1 | XP_016866175.1 | 2 | 278 | |||
XM_017010687.1 | XP_016866176.1 | 3 | 495 | |||
XM_017010688.1 | XP_016866177.1 | 3 | 503 | |||
GenBank mRNA | AK074125.1 | - | 2 | 207 | ||
AK093571.1 | - | 2 | 212 | |||
AK303562.1 | - | 3 | 414 | |||
BC033286.1 | - | 3 | 194 | 4509724 | ||
CR975586.1 | - | 2 | 57 | |||
DC410126.1 | - | 3 | 414 |