Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080451.1 | NP_001073920.1 | 2 | 481 | ||
GenBank mRNA | BC146962.1 | - | 2 | 445 | 9056890 | |
BX248259.1 | - | 2 | 477 |