Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143917.1 | NP_001137389.1 | 2 | 259 | ||
NM_153374.2 | NP_699205.1 | 2 | 850 | |||
XM_017022057.1 | XP_016877546.1 | 2 | 242 | |||
GenBank mRNA | AK129759.1 | - | 2 | 329 | ||
AY125955.1 | - | 2 | 839 | |||
BC022075.1 | - | 2 | 395 | 4799858 | ||
BC033515.1 | - | 2 | 847 | 5175226 |