Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284292.1 | NP_001271221.1 | 3 | 880 | ||
NM_001284293.1 | NP_001271222.1 | 2 | 786 | |||
NM_175741.2 | NP_786883.1 | 3 | 965 | |||
GenBank mRNA | AF482429.1 | - | 2 | 569 | ||
AK098568.1 | - | 2 | 443 | |||
AK302656.1 | - | 3 | 880 | |||
AK302680.1 | - | 2 | 523 | |||
BC114518.1 | - | 2 | 541 | 40036588 | ||
BC114567.1 | - | 2 | 536 | 40036589 |