Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304964.1 | NP_001291893.1 | 3 | 589 | ||
NM_152779.3 | NP_689992.1 | 3 | 589 | |||
XM_011538134.2 | XP_011536436.2 | 3 | 3384 | |||
GenBank mRNA | AY358731.1 | - | 3 | 589 | ||
BC014603.1 | - | 3 | 550 | 4822995 |