Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP7D2 SIRNA Assays ›
Gene Symbol:
MAP7D2
Gene Name:
MAP7 domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.X: 20006713 - 20116996 on Build GRCh38
UniGene ID:
Hs.127951
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s48813
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP7D2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001168465.1
NP_001161937.1
16
2369
NM_001168466.1
NP_001161938.1
14
2111
NM_001168467.1
NP_001161939.1
15
2083
NM_152780.3
NP_689993.2
15
2246
XM_005274478.2
XP_005274535.1
16
2232
XM_005274479.2
XP_005274536.1
16
2199
XM_005274480.3
XP_005274537.1
15
2133
XM_006724481.2
XP_006724544.1
17
2379
XM_006724482.2
XP_006724545.1
16
2256
XM_011545483.1
XP_011543785.1
17
2346
XM_011545484.1
XP_011543786.1
17
2329
XM_011545485.1
XP_011543787.1
17
2264
XM_017029393.1
XP_016884882.1
16
2223
XM_017029394.1
XP_016884883.1
16
2206
XM_017029395.1
XP_016884884.1
16
2182
XM_017029396.1
XP_016884885.1
16
2141
XM_017029397.1
XP_016884886.1
16
2117
GenBank mRNA
AK027409.1
-
15
2148
AK127056.1
-
13
1785
AK294788.1
-
15
2077
AK296319.1
-
14
2022
AK303019.1
-
10
1357
AK316117.1
-
14
2019
BC037165.2
-
13
1913
BC089400.1
-
16
2290
3063898
BC136379.1
-
15
2163
9020366
KP849473.1
-
16
2204
KP849474.1
-
16
2171
KP979782.1
-
16
2238
KP979783.1
-
16
2205
KP979784.1
-
16
2262
KP979785.1
-
16
2229
KP979786.1
-
16
2262
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
structural molecule activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline