Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321395.1 | NP_001308324.1 | 9 | 912 | ||
NM_152781.3 | NP_689994.2 | 9 | 1032 | |||
NR_135630.1 | 9 | 1076 | ||||
XM_005278233.2 | XP_005278290.1 | 10 | 991 | |||
XM_005278235.1 | XP_005278292.1 | 10 | 876 | |||
XM_006721802.3 | XP_006721865.1 | 6 | 514 | |||
XM_011524586.2 | XP_011522888.1 | 9 | 975 | |||
XM_017024424.1 | XP_016879913.1 | 9 | 1012 | |||
GenBank mRNA | AK057392.1 | - | 9 | 1032 | ||
AK301774.1 | - | 10 | 876 | |||
AK301884.1 | - | 10 | 996 | |||
AK301943.1 | - | 9 | 892 | |||
BC033734.1 | - | 8 | 1666 | |||
BC132681.1 | - | 9 | 968 | 40146703 |