Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030019.1 | NP_001025190.1 | 8 | 965 | ||
NM_001284350.1 | NP_001271279.1 | 9 | 1063 | |||
NM_152782.3 | NP_689995.3 | 9 | 881 | |||
XM_017011930.1 | XP_016867419.1 | 9 | 871 | |||
GenBank mRNA | AF429967.1 | - | 8 | 956 | ||
AK302011.1 | - | 9 | 992 | |||
BC026189.2 | - | 9 | 869 | 4819018 | ||
DQ099386.1 | - | 5 | 484 |