Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152784.3 | NP_689997.3 | 7 | 568 | ||
XM_011527885.2 | XP_011526187.1 | 7 | 860 | |||
XM_011527886.2 | XP_011526188.1 | 7 | 860 | |||
XM_011527887.2 | XP_011526189.1 | 7 | 860 | |||
XM_011527888.2 | XP_011526190.1 | 7 | 860 | |||
XM_011527889.2 | XP_011526191.1 | 7 | 860 | |||
XM_011527890.2 | XP_011526192.1 | 7 | 860 | |||
XM_017026564.1 | XP_016882053.1 | 7 | 860 | |||
XM_017026565.1 | XP_016882054.1 | 6 | 513 | |||
XM_017026566.1 | XP_016882055.1 | 7 | 860 | |||
GenBank mRNA | AK128088.1 | - | 6 | 513 | ||
BC043005.1 | - | 7 | 554 | 5297674 |