Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025598.1 | NP_001020769.1 | 5, 6 | 885 | ||
NM_001287600.1 | NP_001274529.1 | 4, 5 | 740 | |||
NM_181720.2 | NP_859071.2 | 5, 6 | 885 | |||
XM_005245070.2 | XP_005245127.1 | 5, 6 | 885 | |||
XM_005245073.3 | XP_005245130.1 | 3, 4 | 638 | |||
XM_011509391.2 | XP_011507693.1 | 4, 5 | 716 | |||
XM_017000960.1 | XP_016856449.1 | 5, 6 | 885 | |||
GenBank mRNA | AK126116.1 | - | 5, 6 | 880 | ||
AK160366.1 | - | 5, 6 | 860 | |||
AL832852.1 | - | 3, 4 | 320 | |||
BC043387.1 | - | 4, 5 | 573 | |||
BC053688.1 | - | 5, 6 | 581 | 5213184 | ||
BX537846.1 | - | 5, 6 | 785 |