Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025598.1 | NP_001020769.1 | 7 | 1166 | ||
NM_001287600.1 | NP_001274529.1 | 6 | 1021 | |||
NM_001287602.1 | NP_001274531.1 | 3 | 635 | |||
NM_181720.2 | NP_859071.2 | 7 | 1166 | |||
XM_005245070.2 | XP_005245127.1 | 7 | 1166 | |||
XM_005245073.3 | XP_005245130.1 | 5 | 919 | |||
XM_011509391.2 | XP_011507693.1 | 6 | 997 | |||
XM_017000960.1 | XP_016856449.1 | 7 | 1166 | |||
GenBank mRNA | AK126116.1 | - | 7 | 1161 | ||
AK126163.1 | - | 3 | 635 | |||
AK160366.1 | - | 7 | 1141 | |||
AL832852.1 | - | 5 | 601 | |||
BC043387.1 | - | 6 | 854 | |||
BC053688.1 | - | 7 | 862 | 5213184 | ||
BX537846.1 | - | 6 | 1526 |