Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025598.1 | NP_001020769.1 | 4 | 710 | ||
NM_001287600.1 | NP_001274529.1 | 3 | 565 | |||
NM_181720.2 | NP_859071.2 | 4 | 710 | |||
XM_005245070.2 | XP_005245127.1 | 4 | 710 | |||
XM_005245073.3 | XP_005245130.1 | 2 | 463 | |||
XM_011509391.2 | XP_011507693.1 | 3 | 541 | |||
XM_017000960.1 | XP_016856449.1 | 4 | 710 | |||
GenBank mRNA | AK126116.1 | - | 4 | 705 | ||
AK160366.1 | - | 4 | 685 | |||
AL832852.1 | - | 2 | 145 | |||
BC043387.1 | - | 3 | 398 | |||
BC053688.1 | - | 4 | 406 | 5213184 | ||
BX537846.1 | - | 4 | 610 |