Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCSAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190259.1 | NP_001177188.1 | 6 | 453 | ||
NM_001190260.1 | NP_001177189.1 | 5 | 402 | |||
NM_152785.4 | NP_689998.1 | 6 | 447 | |||
XM_005247272.2 | XP_005247329.1 | 6 | 871 | |||
XM_011512627.2 | XP_011510929.1 | 6 | 325 | |||
XM_011512628.2 | XP_011510930.1 | 6 | 382 | |||
XM_011512629.1 | XP_011510931.1 | 6 | 874 | |||
XM_011512630.1 | XP_011510932.1 | 6 | 880 | |||
GenBank mRNA | AF521911.1 | - | 6 | 434 | ||
AK313368.1 | - | 6 | 394 | |||
AY212246.1 | - | 6 | 368 | |||
BC030506.1 | - | 6 | 315 | 4385178 | ||
BM456595.1 | - | 6 | 403 | |||
BP224611.1 | - | 5 | 350 | |||
DA455714.1 | - | 6 | 448 |