Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKS4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145865.2 | NP_665872.2 | 1 | 120 | ||
GenBank mRNA | AK096138.1 | - | 1 | 120 | ||
BC111784.1 | - | 1 | 122 | |||
BC112368.1 | - | 1 | 118 |