Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000401.3 | NP_000392.3 | 8 | 1356 | ||
NM_207122.1 | NP_997005.1 | 8 | 1535 | |||
XM_011519950.1 | XP_011518252.1 | 9 | 1354 | |||
XM_011519951.1 | XP_011518253.1 | 9 | 1464 | |||
GenBank mRNA | AK296713.1 | - | 8 | 1049 | ||
AK312375.1 | - | 8 | 1412 | |||
BC010058.1 | - | 8 | 1409 | 3947503 | ||
BC068545.1 | - | 8 | 1375 | 5263866 | ||
BX648142.1 | - | 8 | 1333 | |||
U62740.1 | - | 8 | 1535 | |||
U64511.1 | - | 7 | 1227 |