Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206846.1 | NP_001193775.1 | 6 | 2640 | ||
NM_018136.4 | NP_060606.3 | 6 | 2640 | |||
GenBank mRNA | AF509326.1 | - | 6 | 2383 | ||
AK001380.1 | - | 2 | 315 | |||
AK125107.1 | - | 6 | 2640 | |||
AK226178.1 | - | 6 | 2573 | |||
AY099891.1 | - | 4 | 1165 | |||
AY099893.1 | - | 4 | 1165 | |||
AY367065.1 | - | 6 | 2383 | |||
AY971956.1 | - | 6 | 2383 | |||
BC034607.1 | - | 2 | 259 |