Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152901.3 | NP_690865.1 | 2 | 522 | ||
GenBank mRNA | AF086332.1 | - | 2 | 522 | ||
AF454669.1 | - | 2 | 511 | |||
AF467809.1 | - | 2 | 517 | |||
AY163727.1 | - | 2 | 522 |