Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033025.2 | NP_001028197.1 | 5 | 1270 | ||
NM_001261440.1 | NP_001248369.1 | 5 | 1267 | |||
NM_001261441.1 | NP_001248370.1 | 6 | 1294 | |||
NM_001261442.1 | NP_001248371.1 | 4 | 1235 | |||
NM_001439.3 | NP_001430.1 | 5 | 1306 | |||
NR_048570.1 | 6 | 1401 | ||||
XM_005270621.1 | XP_005270678.1 | 6 | 1136 | |||
XM_011540994.2 | XP_011539296.1 | 6 | 1562 | |||
XM_011540995.2 | XP_011539297.1 | 7 | 1579 | |||
XM_011540996.2 | XP_011539298.1 | 6 | 781 | |||
XM_017000651.1 | XP_016856140.1 | 5 | 907 | |||
GenBank mRNA | AB009284.1 | - | 5 | 939 | ||
AF000416.1 | - | 5 | 939 | |||
AK096914.1 | - | 5 | 900 | |||
AK298124.1 | - | 6 | 1054 | |||
AK312895.1 | - | 5 | 875 | |||
AU130571.1 | - | 6 | 779 | |||
BC014910.1 | - | 5 | 826 | |||
BC036015.1 | - | 5 | 916 | 5273246 | ||
BC045681.1 | - | 5 | 2089 | 5300220 | ||
U76189.1 | - | 1 | 49 |