Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153757.3 | NP_715638.1 | 1 | 1101 | ||
GenBank mRNA | AK054689.1 | - | 1 | 695 | ||
AL834329.1 | - | 1 | 678 | |||
BC039416.1 | - | 1 | 71 | |||
BC104883.1 | - | 1 | 644 | 8143886 |