Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172006.2 | NP_742003.1 | 4 | 351 | ||
NM_172131.2 | NP_742143.1 | 3 | 196 | |||
XM_017027824.1 | XP_016883313.1 | 5 | 423 | |||
XM_017027825.1 | XP_016883314.1 | 5 | 676 | |||
GenBank mRNA | AF454506.1 | - | 4 | 351 | ||
AF454507.1 | - | 3 | 196 | |||
BC121071.1 | - | 4 | 349 | 40119593 | ||
BC121072.1 | - | 4 | 349 | 40119597 |