Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR51B5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005567.2 | NP_001005567.2 | 5 | 1300 | ||
GenBank mRNA | BC136707.1 | - | 1 | 911 | 9020697 | |
BC136708.1 | - | 1 | 911 | 9020698 |