Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010877.3 | NP_001010877.2 | 6 | 1049 | ||
NM_001318534.1 | NP_001305463.1 | 6 | 1095 | |||
NM_001318535.1 | NP_001305464.1 | 6 | 1068 | |||
XM_005249019.3 | XP_005249076.1 | 7 | 1754 | |||
XM_006715067.2 | XP_006715130.1 | 7 | 1903 | |||
XM_011514510.1 | XP_011512812.1 | 7 | 2591 | |||
XM_011514511.2 | XP_011512813.1 | 8 | 2337 | |||
XM_011514512.1 | XP_011512814.1 | 7 | 2567 | |||
XM_011514513.1 | XP_011512815.1 | 7 | 2564 | |||
XM_011514514.1 | XP_011512816.1 | 7 | 2516 | |||
XM_011514516.1 | XP_011512818.1 | 6 | 1559 | |||
XM_011514517.1 | XP_011512819.1 | 7 | 2538 | |||
XM_011514518.1 | XP_011512820.1 | 7 | 2511 | |||
XM_017010756.1 | XP_016866245.1 | 7 | 1364 | |||
XM_017010757.1 | XP_016866246.1 | 7 | 1214 | |||
XM_017010758.1 | XP_016866247.1 | 7 | 2492 | |||
XM_017010759.1 | XP_016866248.1 | 8 | 1401 | |||
XM_017010760.1 | XP_016866249.1 | 7 | 1764 | |||
XM_017010761.1 | XP_016866250.1 | 6 | 2146 | |||
GenBank mRNA | AK289881.1 | - | 7 | 1227 | ||
AL833166.1 | - | 6 | 1749 | |||
BC136745.1 | - | 6 | 1110 | 9020735 | ||
BC144385.1 | - | 6 | 1156 | |||
BC144386.1 | - | 6 | 1129 |