Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010877.3 | NP_001010877.2 | 6 | 1805 | ||
NM_001318534.1 | NP_001305463.1 | 6 | 1851 | |||
NM_001318535.1 | NP_001305464.1 | 6 | 1824 | |||
XM_005249019.3 | XP_005249076.1 | 7 | 2510 | |||
XM_006715067.2 | XP_006715130.1 | 7 | 2659 | |||
XM_011514510.1 | XP_011512812.1 | 7 | 3347 | |||
XM_011514511.2 | XP_011512813.1 | 8 | 3093 | |||
XM_011514512.1 | XP_011512814.1 | 7 | 3323 | |||
XM_011514513.1 | XP_011512815.1 | 7 | 3320 | |||
XM_011514514.1 | XP_011512816.1 | 7 | 3272 | |||
XM_011514516.1 | XP_011512818.1 | 6 | 2315 | |||
XM_011514517.1 | XP_011512819.1 | 7 | 3294 | |||
XM_011514518.1 | XP_011512820.1 | 7 | 3267 | |||
XM_017010756.1 | XP_016866245.1 | 7 | 2120 | |||
XM_017010757.1 | XP_016866246.1 | 7 | 1970 | |||
XM_017010758.1 | XP_016866247.1 | 7 | 3248 | |||
XM_017010759.1 | XP_016866248.1 | 8 | 2157 | |||
XM_017010760.1 | XP_016866249.1 | 7 | 2520 | |||
XM_017010761.1 | XP_016866250.1 | 6 | 2902 | |||
GenBank mRNA | AK289881.1 | - | 7 | 1983 | ||
AL833166.1 | - | 6 | 2505 | |||
BC136745.1 | - | 6 | 1866 | 9020735 | ||
BC144385.1 | - | 6 | 1912 | |||
BC144386.1 | - | 6 | 1885 |
Gene Symbol: ZNF33B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF222749.1 | - | 1 | 49 |