Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042427.1 | NP_001035892.1 | 2 | 202 | ||
NM_182554.2 | NP_872360.2 | 2 | 202 | |||
GenBank mRNA | AK091773.1 | - | 2 | 189 | ||
BC028127.1 | - | 2 | 154 | 5164374 | ||
DB043416.1 | - | 2 | 175 |