Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005244.4 | NP_005235.3 | 2 | 423 | ||
NM_172110.3 | NP_742108.2 | 2 | 423 | |||
XM_005260327.2 | XP_005260384.1 | 2 | 116 | |||
XM_017027721.1 | XP_016883210.1 | 2 | 141 | |||
GenBank mRNA | AF387364.1 | - | 2 | 461 | ||
AK293032.1 | - | 2 | 127 | |||
AY705349.1 | - | 1 | 59 | |||
BC008803.2 | - | 2 | 177 | 3944547 | ||
BC013882.2 | - | 2 | 423 | |||
BT006682.1 | - | 1 | 49 | |||
U71207.1 | - | 1 | 51 |