Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM49B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206626.1 | NP_001193555.1 | 1 | 177 | ||
XM_017017561.1 | XP_016873050.1 | 1 | 181 | |||
GenBank mRNA | JF968463.1 | - | 1 | 63 |