Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282560.1 | NP_001269489.1 | 9 | 888 | ||
NM_001282561.1 | NP_001269490.1 | 9 | 888 | |||
NM_001282562.1 | NP_001269491.1 | 9 | 946 | |||
NM_001990.3 | NP_001981.2 | 10 | 1026 | |||
NR_104214.1 | 10 | 1026 | ||||
XM_006710449.3 | XP_006710512.1 | 11 | 1062 | |||
XM_011540999.2 | XP_011539301.1 | 11 | 1065 | |||
XM_011541001.2 | XP_011539303.1 | 10 | 1023 | |||
XM_011541003.2 | XP_011539305.1 | 10 | 925 | |||
XM_011541004.2 | XP_011539306.1 | 10 | 922 | |||
XM_017000652.1 | XP_016856141.1 | 11 | 1066 | |||
XM_017000653.1 | XP_016856142.1 | 12 | 1186 | |||
XM_017000654.1 | XP_016856143.1 | 10 | 1023 | |||
XM_017000655.1 | XP_016856144.1 | 11 | 1179 | |||
XM_017000656.1 | XP_016856145.1 | 10 | 926 | |||
XM_017000657.1 | XP_016856146.1 | 9 | 918 | |||
XM_017000658.1 | XP_016856147.1 | 9 | 943 | |||
XM_017000659.1 | XP_016856148.1 | 8 | 802 | |||
GenBank mRNA | AK289805.1 | - | 10 | 985 | ||
AK295745.1 | - | 9 | 882 | |||
AK298129.1 | - | 9 | 943 | |||
AK303664.1 | - | 8 | 802 | |||
BC029500.1 | - | 10 | 986 | 5272319 | ||
BC041667.1 | - | 9 | 815 | 5743906 | ||
BX648945.1 | - | 10 | 995 | |||
U81602.1 | - | 5 | 625 |