Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CRACR2B SIRNA Assays ›
Gene Symbol:
CRACR2B
Gene Name:
calcium release activated channel regulator 2B
Gene Aliases:
EFCAB4A
Chromosome Location:
Chr.11: 825558 - 831991 on Build GRCh38
UniGene ID:
Hs.660936
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s49152
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CRACR2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286606.1
NP_001273535.1
3
756
NM_173584.4
NP_775855.3
3
756
XM_011520011.1
XP_011518313.1
3
1320
XM_011520019.1
XP_011518321.1
3
1320
XM_017017581.1
XP_016873070.1
3
1320
XM_017017582.1
XP_016873071.1
4
1127
XM_017017583.1
XP_016873072.1
4
1099
XM_017017584.1
XP_016873073.1
4
1139
XM_017017585.1
XP_016873074.1
4
1678
XM_017017586.1
XP_016873075.1
4
999
XM_017017587.1
XP_016873076.1
4
1370
XM_017017588.1
XP_016873077.1
4
794
XM_017017589.1
XP_016873078.1
3
1320
XM_017017590.1
XP_016873079.1
4
909
XM_017017591.1
XP_016873080.1
4
879
XM_017017592.1
XP_016873081.1
4
1458
XM_017017593.1
XP_016873082.1
4
779
XM_017017594.1
XP_016873083.1
4
1150
XM_017017595.1
XP_016873084.1
4
574
XM_017017596.1
XP_016873085.1
3
1100
XM_017017597.1
XP_016873086.1
3
1320
XM_017017598.1
XP_016873087.1
3
1320
XM_017017599.1
XP_016873088.1
4
995
GenBank mRNA
AK075180.1
-
1
194
AK097334.1
-
1
184
AK127926.1
-
1
186
BC033196.1
-
3
537
4849453
BC073821.1
-
4
850
5952348
DA967563.1
-
1
184
GU952799.1
-
3
408
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
store-operated calcium entry
cellular protein localization
regulation of store-operated calcium entry
calcium ion binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9bfc7:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline