Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042463.2 | NP_001035928.2 | 3 | 355 | ||
NM_001276274.1 | NP_001263203.1 | 3 | 355 | |||
NM_174940.3 | NP_777600.3 | 3 | 331 | |||
XM_006718206.3 | XP_006718269.3 | 3 | 361 | |||
XM_017017600.1 | XP_016873089.1 | 3 | 536 | |||
GenBank mRNA | AK124309.1 | - | 2 | 1397 | ||
AK302585.1 | - | 3 | 150 | |||
BC008671.1 | - | 3 | 279 | 3854028 | ||
BI916171.1 | - | 3 | 333 | |||
CF134831.1 | - | 2 | 121 | |||
CK819789.1 | - | 3 | 145 | |||
DR006597.1 | - | 3 | 298 |