Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC88B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032251.5 | NP_115627.6 | 15 | 2765 | ||
XM_006718519.3 | XP_006718582.1 | 15 | 2777 | |||
XM_011544957.2 | XP_011543259.1 | 15 | 2778 | |||
XM_011544959.2 | XP_011543261.1 | 15 | 2778 | |||
XM_011544960.2 | XP_011543262.1 | 15 | 2777 | |||
XM_011544962.2 | XP_011543264.1 | 2 | 400 | |||
XM_017017601.1 | XP_016873090.1 | 15 | 2779 | |||
GenBank mRNA | AK074126.1 | - | 9 | 2199 | ||
AK090436.1 | - | 15 | 2760 | |||
AK095289.1 | - | 6 | 1736 | |||
BC040232.1 | - | 2 | 322 | |||
BC140822.1 | - | 15 | 2754 | 9021690 | ||
BC140824.1 | - | 15 | 2754 | 9021692 | ||
BC141866.1 | - | 15 | 2744 |