Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173591.3 | NP_775862.3 | 58 | 6932 | ||
XM_005268802.3 | XP_005268859.1 | 62 | 7733 | |||
XM_011538191.1 | XP_011536493.1 | 59 | 7009 | |||
XM_011538192.2 | XP_011536494.1 | 56 | 6950 | |||
XM_011538193.1 | XP_011536495.1 | 56 | 6869 | |||
GenBank mRNA | AK075060.1 | - | 6 | 535 | ||
AK096852.1 | - | 12 | 1210 | |||
BC101016.2 | - | 13 | 1167 | |||
BC101017.2 | - | 12 | 1081 | |||
BC101018.1 | - | 12 | 1081 |