Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036555.1 | 3 | 592 | |||
GenBank mRNA | AK091129.1 | - | 3 | 558 | ||
BC101220.1 | - | 3 | 409 | 40022214 | ||
BC101222.2 | - | 3 | 409 | 40022216 | ||
BC101223.1 | - | 3 | 409 | 40022217 |