Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303130.1 | NP_001290059.1 | 9 | 1256 | ||
NM_001303131.1 | NP_001290060.1 | 10 | 1743 | |||
NM_174942.2 | NP_777602.1 | 10 | 1377 | |||
XM_005268804.3 | XP_005268861.1 | 11 | 1446 | |||
XM_011538219.2 | XP_011536521.1 | 10 | 1302 | |||
XM_011538220.2 | XP_011536522.1 | 9 | 2637 | |||
XM_011538221.2 | XP_011536523.1 | 9 | 1487 | |||
XM_017019206.1 | XP_016874695.1 | 11 | 1520 | |||
GenBank mRNA | AK095594.1 | - | 10 | 1743 | ||
AK315189.1 | - | 9 | 1234 | |||
BC043366.1 | - | 10 | 1327 | 5104644 | ||
BX649059.1 | - | 1 | 192 |