Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176818.2 | NP_789788.1 | 3 | 349 | ||
NR_033684.1 | 4 | 447 | ||||
GenBank mRNA | AK094319.1 | - | 3 | 343 | ||
BC034962.1 | - | 4 | 418 | |||
BC047778.1 | - | 4 | 418 | |||
BC064523.1 | - | 3 | 320 | |||
BC107145.1 | - | 3 | 306 | 40031296 | ||
BC107146.1 | - | 3 | 299 |