Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178096.1 | NP_001171567.1 | 4 | 713 | ||
NM_001993.4 | NP_001984.1 | 4 | 713 | |||
GenBank mRNA | AF487337.1 | - | 4 | 493 | ||
AF497569.1 | - | 4 | 440 | |||
AF497570.1 | - | 4 | 440 | |||
AF497571.1 | - | 2 | 213 | |||
AK312638.1 | - | 4 | 609 | |||
AY940730.1 | - | 4 | 493 | |||
AY940731.1 | - | 4 | 493 | |||
BC011029.1 | - | 4 | 660 | 4073183 | ||
BT019808.1 | - | 4 | 492 | |||
CR541792.1 | - | 4 | 492 | |||
J02931.1 | - | 4 | 603 | |||
M16553.1 | - | 4 | 567 | |||
M27436.1 | - | 4 | 591 |