Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRTG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173814.5 | NP_776175.2 | 2 | 351 | ||
XM_011521459.1 | XP_011519761.1 | 2 | 881 | |||
XM_011521460.1 | XP_011519762.1 | 2 | 881 | |||
XM_017022081.1 | XP_016877570.1 | 2 | 438 | |||
XM_017022082.1 | XP_016877571.1 | 2 | 881 | |||
GenBank mRNA | AY630258.1 | - | 2 | 211 | ||
CN352629.1 | - | 2 | 320 | |||
CN352630.1 | - | 2 | 351 |