Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008226.1 | NP_001008227.1 | 3 | 644 | ||
GenBank mRNA | AK304339.1 | - | 4 | 756 | ||
AL833762.1 | - | 3 | 652 | |||
BC144647.1 | - | 3 | 866 |