Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM98B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173611.3 | NP_775882.2 | 6 | 726 | ||
GenBank mRNA | AK095745.1 | - | 6 | 700 | ||
BC035600.1 | - | 5 | 584 | |||
BC093898.1 | - | 6 | 696 | 7939743 |