Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004060.1 | NP_001004060.1 | 12 | 1318 | ||
NM_173614.2 | NP_775885.1 | 12 | 1318 | |||
XM_017023156.1 | XP_016878645.1 | 12 | 1365 | |||
GenBank mRNA | AK225803.1 | - | 12 | 1288 | ||
AL832855.1 | - | 12 | 1312 | |||
BC028389.1 | - | 11 | 1178 | 4824724 | ||
BC041131.1 | - | 12 | 1292 | 5757234 |
Gene Symbol: NOMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ954152.1 | - | 583 |