Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004060.1 | NP_001004060.1 | 8, 9 | 943 | ||
NM_173614.2 | NP_775885.1 | 8, 9 | 943 | |||
XM_017023156.1 | XP_016878645.1 | 8, 9 | 990 | |||
GenBank mRNA | AK225803.1 | - | 8, 9 | 913 | ||
AL832855.1 | - | 8, 9 | 937 | |||
BC028389.1 | - | 7, 8 | 803 | 4824724 | ||
BC041131.1 | - | 8, 9 | 917 | 5757234 |
Gene Symbol: NOMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014287.3 | NP_055102.3 | 8, 9 | 937 | ||
GenBank mRNA | BC065535.1 | - | 8, 9 | 890 | 6007603 | |
X57398.1 | - | 8, 9 | 775 |
Gene Symbol: NOMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004067.3 | NP_001004067.1 | 8, 9 | 1006 | ||
XM_005255318.1 | XP_005255375.1 | 8, 9 | 1000 | |||
GenBank mRNA | AU132793.1 | - | 1, 2 | 68 | ||
BQ954152.1 | - | 208 | ||||
CD653707.1 | - | 2, 3 | 292 | |||
DA392676.1 | - | 3, 4 | 335 |