Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CES4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173815.6 | NP_776176.5 | 2 | 270 | ||
XM_005255896.4 | XP_005255953.1 | 2 | 267 | |||
XM_011523021.2 | XP_011521323.1 | 2 | 267 | |||
XM_011523023.2 | XP_011521325.1 | 2 | 267 | |||
XM_011523024.2 | XP_011521326.1 | 2 | 267 | |||
XM_011523025.2 | XP_011521327.1 | 2 | 267 | |||
XM_011523026.2 | XP_011521328.1 | 2 | 267 | |||
XM_011523027.2 | XP_011521329.1 | 2 | 267 | |||
XM_011523028.2 | XP_011521330.1 | 2 | 275 | |||
XM_011523029.2 | XP_011521331.1 | 2 | 267 | |||
XM_011523030.2 | XP_011521332.1 | 2 | 267 | |||
XM_017023160.1 | XP_016878649.1 | 2 | 267 | |||
XM_017023161.1 | XP_016878650.1 | 2 | 267 | |||
XM_017023162.1 | XP_016878651.1 | 2 | 267 | |||
XM_017023163.1 | XP_016878652.1 | 2 | 267 | |||
XM_017023164.1 | XP_016878653.1 | 2 | 267 | |||
XM_017023165.1 | XP_016878654.1 | 2 | 267 | |||
XM_017023166.1 | XP_016878655.1 | 2 | 267 | |||
GenBank mRNA | AK296064.1 | - | 2 | 190 | ||
DC339496.1 | - | 2 | 270 |