Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK295350.1 | - | 1 | 56 |
Gene Symbol: LINC00921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033904.1 | 1 | 88 | |||
GenBank mRNA | AK096958.1 | - | 1 | 54 | ||
BC104941.1 | - | 1 | 113 | |||
BC104943.1 | - | 1 | 113 | |||
DA532447.1 | - | 1 | 90 |