Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSEN54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207346.2 | NP_997229.2 | 10, 11 | 1456 | ||
XM_005257229.3 | XP_005257286.1 | 10, 11 | 1643 | |||
GenBank mRNA | BC045551.1 | - | 11, 12 | 1383 | ||
BC047793.1 | - | 10, 11 | 1425 | 6500549 | ||
BC053643.1 | - | 10, 11 | 1427 | |||
BE251582.1 | - | 3, 4 | 226 |