Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC57 SIRNA Assays ›
Gene Symbol:
CCDC57
Gene Name:
coiled-coil domain containing 57
Gene Aliases:
-
Chromosome Location:
Chr.17: 82101458 - 82212892 on Build GRCh38
UniGene ID:
Hs.631724
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s49439
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC57
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001316321.1
NP_001303250.1
5, 6
997
NM_198082.2
NP_932348.2
4, 5
663
XM_011523545.2
XP_011521847.2
5, 6
900
XM_011523546.2
XP_011521848.1
4, 5
1232
XM_011523548.2
XP_011521850.1
5, 6
928
XM_011523549.2
XP_011521851.1
4, 5
742
XM_011523551.2
XP_011521853.1
5, 6
941
XM_011523552.2
XP_011521854.1
3, 4
300
XM_011523555.2
XP_011521857.1
2, 3
1300
XM_011523556.2
XP_011521858.2
5, 6
894
XM_017024462.1
XP_016879951.1
5, 6
902
XM_017024463.1
XP_016879952.1
5, 6
891
XM_017024464.1
XP_016879953.1
5, 6
893
XM_017024465.1
XP_016879954.1
5, 6
892
XM_017024466.1
XP_016879955.1
5, 6
881
XM_017024467.1
XP_016879956.1
5, 6
1057
XM_017024468.1
XP_016879957.1
5, 6
888
XM_017024469.1
XP_016879958.1
5, 6
720
XM_017024470.1
XP_016879959.1
4, 5
521
XM_017024471.1
XP_016879960.1
4, 5
697
XM_017024472.1
XP_016879961.1
5, 6
918
XM_017024473.1
XP_016879962.1
5, 6
887
XM_017024474.1
XP_016879963.1
5, 6
891
XM_017024475.1
XP_016879964.1
4, 5
533
XM_017024476.1
XP_016879965.1
4, 5
652
XM_017024477.1
XP_016879966.1
5, 6
820
XM_017024478.1
XP_016879967.1
3, 4
283
XM_017024479.1
XP_016879968.1
5, 6
927
XM_017024480.1
XP_016879969.1
5, 6
900
XM_017024481.1
XP_016879970.1
5, 6
900
XM_017024482.1
XP_016879971.1
5, 6
904
XM_017024483.1
XP_016879972.1
4, 5
548
GenBank mRNA
BC040264.1
-
5, 6
981
5165045
BC110997.1
-
4, 5
663
5725904
BE146089.1
-
2, 3
256
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-nlx55:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline