Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEXDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173620.2 | NP_775891.2 | 10 | 1474 | ||
XM_005256346.3 | XP_005256403.1 | 9 | 1062 | |||
XM_006722280.3 | XP_006722343.1 | 9 | 1037 | |||
XM_011523560.2 | XP_011521862.1 | 8 | 1005 | |||
XM_011523561.2 | XP_011521863.1 | 9 | 1043 | |||
GenBank mRNA | BC035561.1 | - | 10 | 1155 | 5211583 |