Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130918.1 | NP_001124390.1 | 13 | 1972 | ||
NM_173623.3 | NP_775894.2 | 6 | 1060 | |||
XM_017024491.1 | XP_016879980.1 | 13 | 2115 | |||
XM_017024492.1 | XP_016879981.1 | 12 | 2228 | |||
XM_017024493.1 | XP_016879982.1 | 14 | 2182 | |||
XM_017024494.1 | XP_016879983.1 | 13 | 2117 | |||
XM_017024495.1 | XP_016879984.1 | 12 | 2845 | |||
XM_017024496.1 | XP_016879985.1 | 8 | 1255 | |||
XM_017024497.1 | XP_016879986.1 | 13 | 2537 | |||
GenBank mRNA | AK093127.1 | - | 5 | 924 | ||
AK097392.1 | - | 13 | 1884 | |||
AK302952.1 | - | 13 | 1918 | |||
AK310321.1 | - | 6 | 1092 | |||
AL834151.1 | - | 13 | 2538 | |||
BC033563.1 | - | 6 | 1061 | 4825815 | ||
BC041368.2 | - | 13 | 2968 |